Learn More
Eppendorf™ CellXpert™ CS220 - Large Capacity CO2 Incubator Shaker
Accelerate your research with the Eppendorf™ CellXpert™ CS220 CO2 Incubator Shaker - the only CO2 incubator shaker with integrated 180°C sterilization. Achieve high flask capacity and reliable results with optimized lab space efficiency and low operating costs.
$77,668.67
Specifications
| Load Bearing Capacity (Metric) | 18 kg |
|---|---|
| Voltage | 100/130 V |
| Temperature Range (Metric) | 8°C above ambient to 60°C |
| Product Type | Incubator Shaker |
| Temperature Accuracy | ± 0.4°C at 37°C |
| Catalog Number | Mfr. No. | Orbit | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Orbit | Price | Quantity & Availability | |||||
|
01670843
|
Eppendorf™
6740112030 |
51 mm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01670844
|
Eppendorf™
6740111030 |
25 mm |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Accelerate your growth - CellXpert™ CS220 - CO2 Incubator Shaker
The CellXpert™ CS220 is a large capacity, stackable CO2 Incubator Shaker that was designed to accelerate mammalian suspension culture research projects.
Please order separately: platform, clamps, sticky pads, storage base or feet, water supply package. Optional: 1-2 stacking kits and water tank holder
- High flask capacity
- 180°C sterilization routine integrated: including wizard and downloadable performance protocol, no locally restricted UV-airflow/surface-disinfection
- Stainless steel chamber: Hygienic design according to EN ISO 14159:2008 standard plus only few internal parts and no drive wheel, crevices, cables or heating fins inside
- Optimized throughput per lab space: Highest platform size vs. footprint ratio in the market, stackable up to 3-fold
- Saves costs: Always included: slide-out platform, rH-control, touch interface, access port, 180°C sterilization routine, no UV-lamps that require frequent exchange, low gas and energy consumption
- Eppendorf X-drive: Proven and tested shaker drive with four weight-supporting shafts for 24/7 vibration-free shaking
- Glide-up door and slide-out platform: Saves space and ensures fast and easy experiment setup, cleaning and service – even at the back of the chamber
- Full control and fast root-cause analysis: 7 in. touchscreen shows all setpoints and current parameters at a glance, direct visual overview of logged data/events including door openings and alarms, user management
Applications:
- Protein Expression
- Screening
- Cell line development
- Early process development
- Viral vector production in mammalian cells e.g., Ad, AAV, LV
- Bioreactor inoculum preparation
Specifications
| 18 kg | |
| 8°C above ambient to 60°C | |
| ± 0.4°C at 37°C | |
| ± 0.3°C at 37°C | |
| 31.7 in. | |
| 22.8 x 40.4 x 15.2 in. | |
| 522.5 lb. | |
| CO2 Accuracy: ± 0.3% at 5% CO2 | |
| CellXpert CS220 | |
| Active bidirection humidity control up to 85% relative humidity, non-condensing | |
| 224 L |
| 100/130 V | |
| Incubator Shaker | |
| 0.1°C increments | |
| 50 to 60 Hz | |
| 22.8 in. | |
| 20.2 x 36.6 in. | |
| 237 kg | |
| CO2 Range: 0.1% to 20% CO2, CO2 Sensor type: NDIR | |
| Color Touch Screen | |
| Eppendorf X-drive | |
| No |