missing translation for 'onlineSavingsMsg'
Learn More

Medisca Cellulose NF/EP (Microcrystalline)(Flocel 101)

Catalog No. 14185655
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185655 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185655 Supplier Medisca Supplier No. 056708
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cellulose NF / EP is a fine, white to almost white, slightly hygroscopic, powder.

Cellulose NF / EP is a fine, white to almost white, slightly hygroscopic, powder. It consists of free-flowing, nonfibrous particles that are insoluble in water, in dilute acids, and in most organic solvents. Microcrystalline cellulose typically acts as a binder in tablet formation.

Alternative name(s): microcrystalline cellulose

Specifications

Chemical Name or Material Cellulose NF/EP (Microcrystalline)(Flocel 101)
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 9004-34-6
Type Drug Standard
Molecular Weight (g/mol) 160 000 - 560 000
Quantity 500 g
Grade EP, NF