missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose, microcrystalline

CAS: 9004-34-6

Supplier:  Thermo Scientific Chemicals A177300C

 View more versions of this product

Catalog No. AAA177300C

Only null left
Explore available promotions

Description

Description

Cellulose, microcrystalline is used as an emulsifier, bulking agent in food production, extender and anti-caking agent. It is also used in vitamin supplements or tablets. It acts as an alternative to carboxymethylcellulose. It finds application in partition chromatography. Further, it is used as a thickener and stabilizer. In addition to this, it is used as an excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cellulose
Microcrystalline
MFCD00081512
Slightly soluble in sodium hydroxide. Insoluble in water,ethanol,ether and dilute mineral acids.
microcrystalline
9004-34-6
14,1965
10,000 g

Chemical Identifiers

9004-34-6
MFCD00081512
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H335
May cause respiratory irritation.

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

H500

EINECSNumber : 232-674-9

RTECSNumber : FJ5691460

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only