missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose, microcrystalline

Catalog No. p-3346690
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1773036 500 g
AAA177300E 2500 g
AAA177300C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1773036 Supplier Thermo Scientific Chemicals Supplier No. A1773036
Only null left

CAS: 9004-34-6

Cellulose, microcrystalline is used as an emulsifier, bulking agent in food production, extender and anti-caking agent. It is also used in vitamin supplements or tablets. It acts as an alternative to carboxymethylcellulose. It finds application in partition chromatography. Further, it is used as a thickener and stabilizer. In addition to this, it is used as an excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-34-6
MDL Number MFCD00081512

Specifications

Physical Form Microcrystalline
Merck Index 14,1965
Solubility Information Slightly soluble in sodium hydroxide. Insoluble in water,ethanol,ether and dilute mineral acids.
Quantity 500 g
Chemical Name or Material Cellulose, microcrystalline
Hazard Category H500
Hazard Statement GHS H Statement
H335
Precautionary Statement GHS P Statement
P261-P271-P304+P340-P312-P405-P403+P233-P501a
EINECSNumber 232-674-9
RTECSNumber FJ5691460
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only