Learn More
Sartorius Cellulose Extraction Thimbles
Extraction thimbles been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. Designed for versatility, the thimbles can be used with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
Supplier: Sartorius FT1201040100
Description
- Composed of high-purity cellulose for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Chemical Identifiers
| Cellulose | |
| C300 | |
| Extraction Thimbles |
Specifications
| 40 mm | |
| 1.7 mm | |
| 100 mm |
| Cellulose | |
| C300 | |
| Extraction Thimbles |