Learn More
Invitrogen™ CellTrace™ Yellow Cell Proliferation Kit, for flow cytometry
Description
This reagent is specifically designed for excitation by either a 532-nm or 561-nm laser, and it can be easily multiplexed with reagents and antibodies excited by other commonly used lasers.
• Superior performance—bright, single-peak staining enables visualization of multiple generations
• Long-term signal stability—well retained in cells for several days post stain
• Versatile—multiple colors available to easily combine with antibodies or markers of cell function
• Simple—robust staining protocol
Superior fluorescent staining
Successful proliferation analysis by dye dilution requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence after several cell divisions. The intense fluorescent staining provided by CellTrace Yellow dye enables the visualization of six or more generations of proliferating cells before the signal is overwhelmed by intrinsic cellular auto-fluorescence. This kit enables consistent, homogeneous staining results with little fluorescence variation between cells in a population, so distinct generations can be seen without any requirement for complex modeling software.
Long-term signal retention
Unlike stains that label the lipid membrane of cells, CellTrace Yellow stain easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye offers a consistent signal, even after several days in a cell culture environment. CellTrace Yellow dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells.
Easy multiplexing with other fluorophores
The yellow excitation at 546 nm and emission at 579 nm of CellTrace Yellow dye make it ideal for multiplexing due to the limited spectral overlap with other common dyes (e.g., Alexa Fluor™488 and FITC) and green fluorescent protein (GFP) .
Use the SpectraViewer to help you plan your experiments.
Simple, robust staining protocol
The CellTrace Yellow Cell Proliferation Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1 μL of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
Specifications
Specifications
| Content And Storage |
|
| Detection Method | Fluorescence |
| For Use With (Application) | Proliferation Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Cell Proliferation Kit |
| Dye Type | CellTrace™ Yellow |
| Format | Tube(s) |
| Product Line | CellTrace |
| Quantity | 180 Assays |
| Shipping Condition | Room Temperature |
| Show More |
Frequently Asked Questions (FAQs)
Calcein, AM and FDA (fluorescein diaceate) are examples of some dyes used for this application. Since these dyes are not incorporated or covalently attached to any cellular components, they may have a short retention time as some cell types may actively efflux the dye out of the cells. The CellTracker and CellTrace dyes include either a mild thiol-reactive chloromethyl group or amine-reactive succinnimidyl ester group to allow for covalent binding to cellular components, providing for better retention. As with any reagent, one should empirically determine retention times for the cell type used.
Calcein, AM is a good choice for cell tracking and as a general cytoplasmic stain. However, it doesn't bind to anything and may be actively pumped out of the cells within a couple hours, which is likely what happened. The retention of Calcein within live cells is dependent upon the inherent properties of the cell type and culture conditions.
For long-term imaging, you may wish to consider a reactive cytoplasmic stains such as CFDA, SE or the CellTracker and CellTrace dyes.
We have not tested the use of the CellTrace reagents for co-culture applications. In theory, this may work, but you would have to test this on your cells of interest.
A single broad peak is usually caused by using too high a concentration of dye and/or too long an incubation time.
The dyes provided in the CellTrace Proliferation kits are amine-reactive dyes that are considered general cytoplasmic stains. They may bind to various membrane proteins, organelles as well as components in the cytoplasm. They are not known to localize in any specific organelles.
For Research Use Only. Not for use in diagnostic procedures.