missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures
GREENER_CHOICE

Catalog No. 07000727
Change view
Click to view available options
Capacity (Metric):
3.3 mL (Well)
1.7 mL
6.5 mL
16 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
07000727 1.7 mL
07000646 16 mL
07000670 3.3 mL (Well)
07000688 6.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07000727 Supplier Greiner Bio-One Supplier No. 677102
Only null left

Compatible with automated systems. Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures are used for anchorage dependant cell lines.

  • Manufactured from transparent virgin polystyrene
  • Low auto fluorescence
  • Single position lids to prevent cross-contamination
  • Alphanumeric well coding
  • With hydrophobic surface for suspension cultures and hybridoma cell

Service Plan

No

Warranty

No

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 1.7 mL
Color Clear
Culture Area 1 sq. cm
Material Polystyrene
Shape Rectangular
Sterility Sterile
Type Multi-well Cell Culture Plate
Well Count 48
Includes Lid
No. per Case 100
No. per Pack 1
Dimensions (L x W x H) 127.8 x 85.5 x16.5 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm
Show More Show Less