Learn More
Invitrogen™ CellMask™ Plasma Membrane Stains
Description
Invitrogen CellMask™ plasma membrane stains enable fast and uniform labeling of the plasma membrane in almost all cell types. These stains works rapidly and can be detected using standard microscope filter sets with any imaging instrument. Depending upon the cell type and experimental conditions, the plasma membrane staining will last for 60–90 minutes without detectable internalization, providing enough time for most live cell dynamic studies. The stain survives fixation, but not permeabilization.
Features of CellMask™ plasma membrane stains include:
• Sensitivity—specific and rapid plasma membrane staining
• Excellent retention—staining lasts for 60–90 minutes
• Available in most common microscope channels, green, orange, deep red, and near infrared
Drawbacks of other methods
The plasma membrane is a convenient marker of cell boundaries and as such, a number of probes have been used for staining of the membrane. Typically, dyes of a lipophilic nature are used; however, they internalize rapidly, offering a very narrow window for imaging. Fluorescently labeled lectins, such as wheat germ agglutinin, have also been employed as plasma membrane stains. Conjugated lectins depend on cell surface sugars for staining and as a result, stain inconsistently with variation across cell types. Robust plasma membrane staining is important for a range of applications including translocation assays, plasma membrane dynamics, and as a general tool for cell identification in traditional and automated imaging and analysis.
How CellMask™ plasma membrane stains work
CellMask™ plasma membrane stains are designed to deliver uniform staining of the plasma membrane across a wide variety of mammalian cell types and are slow to internalize, particularly compared to traditional approaches such as DiI, DiO, and labeled wheat germ agglutinin.
CellMask™ plasma membrane stains are amphipathic molecules containing a lipophilic moiety for excellent membrane loading and a negatively charged hydrophilic dye for anchoring of the probe in the plasma membrane. While CellMask™ plasma membrane stains provide ample opportunity for live cell imaging, the staining pattern is also maintained after fixation with formaldehyde, enabling additional multiparametric imaging options. However, staining with CellMask™ plasma membrane stains does not survive detergent extraction and, therefore, cannot be used in conjunction with probes that require permeabilization.
Specifications
Specifications
| Color | Green |
| Content And Storage | Store in freezer (–5° to –30°C) and protect from light. |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope |
| Quantity | 100 μL |
| Volume (Metric) | 100 μL |
| Description | CellMask™ Green Plasma Membrane Stain |
| Product Line | Cellmask |
| Label Type | Fluorescent Dye |
| Product Type | Plasma Membrane Stain |
| Sub Cellular Localization | Plasma Membrane |
Frequently Asked Questions (FAQs)
No. For paraffin sections, there are few options due to the delipidation of the membranes by solvents used in the deparaffinization steps. The only good option is to use an antibody against a plasma membrane protein.
Depending upon the cell type and experimental conditions, the plasma membrane staining will last for 60-90 min without detectable internalization, providing enough time for most live cell dynamic studies. The stain survives fixation, but not permeabilization.
The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.
For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).