missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellEvent™ Senescence Green Detection Kit

Catalog No. C10851
Encompass
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10851 100 μL
C10850 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10851 Supplier Invitrogen™ Supplier No. C10851

CellEvent™ Senescence Green Detection Kit consists of a fluorescent probe and optimized buffer that enable the image-based detection of senescent cells

This fluorescein-based probe contains two galactoside moieties, making it a target for β-galactosidase. Activation of β-galactosidase is commonly used as a biomarker for senescent cells. This hydrolase enzyme resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. The enzymatically cleaved product is retained within the cell due to covalent binding of intracellular proteins and emits a fluorogenic signal that has absorption/emission maxima of 490/514 nm.

Kit features include:
• Reliable and quick fluorescent detection of senescent cells
• Greater sensitivity than traditional colorimetric x-gal
• Fluorescent signal is well retained and detected using standard Alexa Fluor™ 488/FITC filter sets
• Multiplex-enabled—fluorescent senescence probe can be multiplexed

Due to a limited replicative lifespan, normal cells enter cell cycle arrest, also known as cellular senescence. While in this senescent phase the cells remain metabolically active without undergoing cell death or division. They adopt a specific phenotypic that includes the appearance of multiple nuclei, increased vacuolization, expression of pH-dependent β-galactosidase, and morphological enlargement and extension. Senescence, through a variety of mechanisms, may play a role in tumor suppression, tumor progression, aging, and tissue repair.


Activation of β-galactosidase is commonly used as a biomarker for senescent cells. This hydrolase enzyme resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. The activity is optimal at lysosomal pH 4, but conventional assays measure at pH 6. It has been shown that normalized β-galactosidase activity is twice as high in senescent cells as in pre-senescent cells regardless of the pH value used for testing.


A colorimetric substrate for β-galactosidase, 5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside (x-gal), has long been used to detect metabolic activity in cells in vitro. However, its use is limited due to inconsistent signal, length of assay, and inability to multiplex.


Therefore, we have developed a sensitive, fluorescent substrate for β-galactosidase that can be used for the detection of senescent cells. The CellEvent™ Senescence Green probe is a fluorescein-based reagent that contains two galactoside moieties, making it a specific target of β-galactosidase. The enzyme- cleaved product is retained within the cell due to covalent binding of intracellular proteins and emits a fluorogenic signal that has absorption/emission maxima of 490/514 nm.

The CellEvent™ Senescent Green Kit provides CellEvent™ Senescent Green probe and an optimized buffer for the detection of senescent cells in fixed samples. This probe can be multiplexed with other fluorescent reagents compatible with paraformaldehyde fixation.

Specifications

Cell Type Mammalian Cells
Content And Storage Store at 2°C to 8°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, High Content Analysis Instrument
Product Type Senescence Green Detection Kit
Format 96-well plate
Product Line CellEvent
Quantity 100 μL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Which sample types can I use with the CellEvent Senescence Green Detection Kit?

The CellEvent Senescence Green Detection Kit can be used with various sample types, including fixed cells, adherent cells, and tissue sections.

Can I use the CellEvent Senescence Green Detection Kit (Cat. Nos. C10850, C10851) on already fixed samples, rather than immediately after fixation?

The CellEvent Senescence Green Detection Kit (C10850) requires fixation for the stain to enter lysosomes. The beta-galactosidase remains active immediately following fixation with paraformaldehyde, so there is a requirement to stain the cells shortly after fixation.

We have not tested fixation and long-term storage. The beta-galactosidase activity could decrease if you wait too long after fixation to stain. Therefore, we do not recommend using the CellEvent Senescence Green Detection Kit on already fixed samples.

Can the CellEvent Senescence Green Probe be multiplexed using antibodies for intracellular labeling?

Yes. After incubating the cells with CellEvent Senescence Green Probe, permeabilize the cells, wash, and proceed with detection.

Can I use my own buffer with the CellEvent Senescence Green Probe?

We do not recommend using your own buffer with the CellEvent Senescence Green Probe. The assay is pH sensitive and we recommend that you use the buffer provided in the kit be used.

Can I use the CellEvent Senescence Green Probe on live cells?

No. It only works on fixed cells; b-galactosidase is still active immediately following fixation with paraformaldehyde.

For Research Use Only. Not for use in diagnostic procedures.