missing translation for 'onlineSavingsMsg'
Learn More

Cell and Molecular Ultra Pure Water, Sterile, Cytiva Hyclone™

Catalog No. WPWCMG1X6
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WPWCMG1X6 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WPWCMG1X6 Supplier Cytiva Supplier No. WPWCMG1X6
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A universal, ultra pure water for your most sensitive research applications.

  • Sterile, extremely low in endotoxin
  • Suitable for cell culture applications, media and reagent preparations
  • Qualified for research investigations of nucleic acid and protein systems
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Purified utilizing a nine-step process resulting in the highest quality water on the market

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Endotoxin <0.25 EU/mL
Grade Cell Culture, Molecular Biology
Packaging PET Plastic Bottle
Sterility Sterile
Certificate of Analysis Yes
Quantity 6 x 1 L
ShelfLife 36 Months
Recommended Storage 2-30°C