missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cell Medium ACF, without glutamine and NaHCO3
GREENER_CHOICE

Catalog No. 670080
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670080 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 670080 Supplier Gibco™ Supplier No. 670080

A complete medium.

A complete animal-origin free medium.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free
Form Solid
Product Type Cell Culture Media
Without Additives No Glutamine
Format Bottle(s)
Quantity 500 g
Source Animal origin-free