missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Cell Culture 1-Tray, 630 cm⊃2

Meticulously engineered cell culture trays suitable for efficient and consistent commercial scale-up for the bioproduction of monoclonal antibodies, extracellular vesicles, vaccines, stem cells, and enzymes.

$322.53 - $3887.80

Specifications

Sterility Sterile
Culture Area 630 cm²
View More Specs

Products 2
Catalog Number Mfr. No. No. per Pack Price Quantity  
Catalog Number Mfr. No. No. per Pack Price Quantity  
MP096143001
View Documents
MP Biomedicals™
096143001
14
Pack of 14 Pair for $3,887.80
Only null left
 
MP096143011
View Documents
MP Biomedicals™
0961430011
1
Pack for $322.53
Only null left
 
Description

Description

  • Graduated on both sides
  • Tissue culture treated surface allows for effective cell adhesion and growth
  • Comes with closed and filtered caps
  • Leak-free
  • Irradiation sterilized
  • Pyrogen-free
  • DNase-free, RNase-free
Specifications

Specifications

Sterile
630 cm²
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.