missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Cell Culture 1-Tray, 630 cm⊃2
Meticulously engineered cell culture trays suitable for efficient and consistent commercial scale-up for the bioproduction of monoclonal antibodies, extracellular vesicles, vaccines, stem cells, and enzymes.
Supplier: MP Biomedicals™ 0961430011
Description
- Graduated on both sides
- Tissue culture treated surface allows for effective cell adhesion and growth
- Comes with closed and filtered caps
- Leak-free
- Irradiation sterilized
- Pyrogen-free
- DNase-free, RNase-free
Specifications
| 1 | |
| Sterile | |
| 630 cm² |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.