missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Celecoxib | |
| 163°C | |
| C17H14F3N3O2S | |
| 1 g | |
| RZEKVGVHFLEQIL-UHFFFAOYSA-N | |
| 4-[5-(4-methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide | |
| 2662 | |
| 381.37 | |
| Crystalline Powder |
| 169590-42-5 | |
| White | |
| MFCD00941298 | |
| celecoxib, celebrex, celebra, onsenal, celocoxib, celecox, xilebao, 4-5-4-methylphenyl-3-trifluoromethyl-1h-pyrazol-1-yl benzenesulfonamide, celecoxib usan, celecoxib old rn | |
| CC1=CC=C(C=C1)C2=CC(=NN2C3=CC=C(C=C3)S(=O)(=O)N)C(F)(F)F | |
| 381.373 | |
| CHEBI:41423 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 169590-42-5 | |
| 381.373 | |
| RZEKVGVHFLEQIL-UHFFFAOYSA-N | |
| 2662 | |
| 4-[5-(4-methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide |
| C17H14F3N3O2S | |
| MFCD00941298 | |
| celecoxib, celebrex, celebra, onsenal, celocoxib, celecox, xilebao, 4-5-4-methylphenyl-3-trifluoromethyl-1h-pyrazol-1-yl benzenesulfonamide, celecoxib usan, celecoxib old rn | |
| CHEBI:41423 | |
| CC1=CC=C(C=C1)C2=CC(=NN2C3=CC=C(C=C3)S(=O)(=O)N)C(F)(F)F |
Safety and Handling
RTECSNumber : DB2944937
TSCA : No