missing translation for 'onlineSavingsMsg'
Learn More

Celecoxib 98.0+%, TCI America™

Catalog No. C2816200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2816200MG 200 mg
C28161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2816200MG Supplier TCI America Supplier No. C2816200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 169590-42-5
Molecular Formula C17H14F3N3O2S
Molecular Weight (g/mol) 381.373
MDL Number MFCD00941298
InChI Key RZEKVGVHFLEQIL-UHFFFAOYSA-N
Synonym celecoxib, celebrex, celebra, onsenal, celocoxib, celecox, xilebao, 4-5-4-methylphenyl-3-trifluoromethyl-1h-pyrazol-1-yl benzenesulfonamide, celecoxib usan, celecoxib old rn
PubChem CID 2662
ChEBI CHEBI:41423
IUPAC Name 4-[5-(4-methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
SMILES CC1=CC=C(C=C1)C2=CC(=NN2C3=CC=C(C=C3)S(=O)(=O)N)C(F)(F)F

Specifications

Melting Point 163°C
Color White
Quantity 200 mg
Formula Weight 381.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Celecoxib
RTECSNumber DB2944937
TSCA No