missing translation for 'onlineSavingsMsg'
Learn More

Celecoxib Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019359
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019359 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019359 Supplier Merck Emd Millipore Supplier No. PHR16831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 169590-42-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H14F3N3O2 S
MDL Number MFCD00941298
Synonym Celecoxib; 4-[5-(4-Methylphenyl)-3-(trifluoromethyl)-1 H-pyrazol-1-yl]benzenesulfonamide; CJ-016377; CP-598107; PF-00345549; PHA-00846533; SC-58635; YM-177
Type Certified Reference Material
Quantity 1 g
Formula Weight 381.37
Grade Certified Reference Material