missing translation for 'onlineSavingsMsg'
Learn More

Ceftriaxone Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012364
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012364 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012364 Supplier Merck Emd Millipore Supplier No. PHR13821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 104376-79-6
Physical Form Neat
Linear Formula C18H16N8 Na2O7S3 · 3.5H2 O
MDL Number MFCD01750405
Synonym Ceftriaxone disodium salt hemi(heptahydrate)
Type Certified Reference Material
Quantity 1 g
Formula Weight 661.60
Grade Certified Reference Material