missing translation for 'onlineSavingsMsg'
Learn More

Ceftizoxime, Thermo Scientific Chemicals

Catalog No. AC465110010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465110010 1 g
AC465110050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465110010 Supplier Thermo Scientific Chemicals Supplier No. 465110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68401-81-0
Molecular Formula C13H13N5O5S2
Molecular Weight (g/mol) 383.40
MDL Number MFCD00072000
InChI Key NNULBSISHYWZJU-LLKWHZGFSA-N
IUPAC Name sodium (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-5-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [H][C@]12SCC=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\C1=CSC(N)=N1)C(O)=O

Specifications

Melting Point 260.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 383.4
Chemical Name or Material Ceftizoxime

RUO – Research Use Only