missing translation for 'onlineSavingsMsg'
Learn More

Ceftiofur Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002506
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002506 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002506 Supplier Merck Emd Millipore Supplier No. PHR15211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ceftiofur Sodium is a caphalosporin antibiotic, with activity similar to other beta-lactam antibiotics. Its mode of action involves the inhibition of the synthesis of bacterial cell wall, thereby leading to cell death.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 104010-37-9
Type Certified Reference Material
Quantity 1 g
Grade Certified Reference Material