missing translation for 'onlineSavingsMsg'
Learn More

Ceftezole 98.0+%, TCI America™

Catalog No. p-7125645
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27705G 5 g
C277025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27705G Supplier TCI America Supplier No. C27705G
Only null left

Chemical Identifiers

CAS 26973-24-0
Molecular Formula C13H12N8O4S3
Molecular Weight (g/mol) 440.471
MDL Number MFCD00274176
InChI Key DZMVCVMFETWNIU-LDYMZIIASA-N
PubChem CID 65755
IUPAC Name (6R,7R)-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-3-(1,3,4-thiadiazol-2-ylsulfanylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)CN3C=NN=N3)C(=O)O)CSC4=NN=CS4

Specifications

Melting Point 163°C
Color Yellow
Quantity 5 g
Formula Weight 440.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ceftezole