accessibility menu, dialog, popup

UserName

CAS RN 26973-24-0

IUPAC Name: sodium (6R,7R)-8-oxo-7-[2-(1H-1,2,3,4-tetrazol-1-yl)acetamido]-3-[(1,3,4-thiadiazol-2-ylsulfanyl)methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: ceftezole
Molecular Weight (g/mol): 462.45
Molecular Formula: C13H11N8NaO4S3
InChi Key: UGUMHWUOXWFPFH-JHQAJZDGSA-M
SMILES: [Na+].[O-]C(=O)C1=C(CSC2=NN=CS2)CS[C@@H]2[C@H](NC(=O)CN3C=NN=N3)C(=O)N12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results