missing translation for 'onlineSavingsMsg'
Learn More

Cefoxitin sodium salt, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111013956
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013956 250 mg
111013954 1 g
111013955 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111013956 Supplier Merck Emd Millipore Supplier No. C4786250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cefoxitin is a β-lactam antibiotic derived from cephamycin C. It is used for intravenous and intramuscular injections. It is found to be active against Bacteroides and is therapeutically used for the treatment of mixed aerobic-anaerobic infections.

Specifications

Packaging Glass Bottle
CAS 33564-30-6
Physical Form Neat
Linear Formula C16H16N3NaO7S2
MDL Number MFCD00079042
Quantity 250 mg
Formula Weight 449.43
Grade Analytical Standard