Learn More
Cefoxitin sodium salt, Analytical Standard, MilliporeSigma™ Supelco™
Cefoxitin is a β-lactam antibiotic derived from cephamycin C. It is used for intravenous and intramuscular injections. It is found to be active against Bacteroides and is therapeutically used for the treatment of mixed aerobic-anaerobic infections.
Supplier: Merck Emd Millipore C47865G
Specifications
| Glass Bottle | |
| Neat | |
| MFCD00079042 | |
| 449.43 |
| 33564-30-6 | |
| C16H16N3NaO7S2 | |
| 5 g | |
| Analytical Standard |