missing translation for 'onlineSavingsMsg'
Learn More

Cefotetan disodium, Thermo Scientific Chemicals

Catalog No. p-7057464
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66206MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66206MC Supplier Thermo Scientific Chemicals Supplier No. J66206MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A broad spectrum antibiotic

A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74356-00-6
Molecular Formula C17H15N7Na2O8S4
Molecular Weight (g/mol) 619.568
MDL Number MFCD00941449
InChI Key ZQQALMSFFARWPK-ZTQQJVKJSA-L
PubChem CID 53024
IUPAC Name disodium;(6R,7S)-7-[[4-(2-amino-1-carboxylato-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)[O-])S4)OC)SC2)C(=O)[O-].[Na+].[Na+]

Specifications

Type Cefotetan diSodium
Physical Form Powder
Color White
Quantity 100 mg
Boiling Point 798°C
Solubility Information Freely soluble in aqueous solution
Formula Weight 619.58
Chemical Name or Material Cefotetan disodium
Hazard Category H315-H317-H319-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 277-834-9
RTECSNumber XI0330600
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only