accessibility menu, dialog, popup

UserName

CAS RN 74356-00-6

IUPAC Name: disodium (6R,7S)-7-{4-[carbamoyl(carboxylato)methylidene]-1,3-dithietane-2-amido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: cefotetan disodium cefotetan sodium
Molecular Weight (g/mol): 619.57
Molecular Formula: C17H15N7Na2O8S4
InChi Key: ZQQALMSFFARWPK-GLHLDKNHSA-L
SMILES: [Na+].[Na+].CO[C@]1(NC(=O)C2SC(S2)=C(C(N)=O)C([O-])=O)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O
Color 
  • (1)
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results