missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefotaxime Sodium Salt 96.0+%, TCI America™
Supplier: TCI America C222425G
Specifications
| Cefotaxime Sodium Salt | |
| 163°C | |
| C16H16N5NaO7S2 | |
| 25 g | |
| AZZMGZXNTDTSME-JUZDKLSSSA-M | |
| sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 88631411 | |
| ≥96.0% (HPLC) |
| 64485-93-4 | |
| White-Yellow | |
| MFCD00079073 | |
| cefotaxime sodium, sodium 6r,7r-3-acetyloxy methyl-7-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetamido-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, cefoperazone sodium salt u.s.p., 6r-6-a,7-b z-3-acetyloxy methyl-7-2-amino-4-thiazolyl methoxyimio acetyl amino-8-oxo-5-thia-1-azabicyclo 4,2,0 oct-2-ene-2-carboxylic acid, sodium salt, sodium 3-acetyloxymethyl-7-2-2-amino-4-thiazolyl-2-methoxyimino-1-oxoethyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, sodium 3-acetyloxymethyl-7-2-2-azanyl-1,3-thiazol-4-yl-2-methoxyimino-ethanoyl amino-8-oxidanylidene-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, sodium 3-acetyloxy methyl-7-2-amino-1,3-thiazol-4-yl methoxyimino acetyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2 | |
| [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 | |
| 477.44 | |
| 477.44 | |
| Crystalline Powder |
Chemical Identifiers
| 64485-93-4 | |
| 477.44 | |
| AZZMGZXNTDTSME-JUZDKLSSSA-M | |
| 88631411 | |
| [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 |
| C16H16N5NaO7S2 | |
| MFCD00079073 | |
| cefotaxime sodium, sodium 6r,7r-3-acetyloxy methyl-7-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetamido-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, cefoperazone sodium salt u.s.p., 6r-6-a,7-b z-3-acetyloxy methyl-7-2-amino-4-thiazolyl methoxyimio acetyl amino-8-oxo-5-thia-1-azabicyclo 4,2,0 oct-2-ene-2-carboxylic acid, sodium salt, sodium 3-acetyloxymethyl-7-2-2-amino-4-thiazolyl-2-methoxyimino-1-oxoethyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, sodium 3-acetyloxymethyl-7-2-2-azanyl-1,3-thiazol-4-yl-2-methoxyimino-ethanoyl amino-8-oxidanylidene-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, sodium 3-acetyloxy methyl-7-2-amino-1,3-thiazol-4-yl methoxyimino acetyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2 | |
| sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Safety and Handling
RTECSNumber : XI0250000
TSCA : No
Recommended Storage : Refrigerator