missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime Sodium Salt 96.0+%, TCI America™

Catalog No. p-7125341
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22245G 5 g
C222425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22245G Supplier TCI America Supplier No. C22245G
Only null left

Chemical Identifiers

CAS 64485-93-4
Molecular Formula C16H16N5NaO7S2
Molecular Weight (g/mol) 477.44
MDL Number MFCD00079073
InChI Key AZZMGZXNTDTSME-JUZDKLSSSA-M
Synonym cefotaxime sodium, sodium 6r,7r-3-acetyloxy methyl-7-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetamido-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, cefoperazone sodium salt u.s.p., 6r-6-a,7-b z-3-acetyloxy methyl-7-2-amino-4-thiazolyl methoxyimio acetyl amino-8-oxo-5-thia-1-azabicyclo 4,2,0 oct-2-ene-2-carboxylic acid, sodium salt, sodium 3-acetyloxymethyl-7-2-2-amino-4-thiazolyl-2-methoxyimino-1-oxoethyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, sodium 3-acetyloxymethyl-7-2-2-azanyl-1,3-thiazol-4-yl-2-methoxyimino-ethanoyl amino-8-oxidanylidene-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate, sodium 3-acetyloxy methyl-7-2-amino-1,3-thiazol-4-yl methoxyimino acetyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2
PubChem CID 88631411
IUPAC Name sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 5 g
Formula Weight 477.44
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cefotaxime Sodium Salt
RTECSNumber XI0250000
TSCA No
Recommended Storage Refrigerator