missing translation for 'onlineSavingsMsg'
Learn More

Cefoperazone sodium salt

Cefoperazone sodium salt, CAS # 62893-20-3, is the sodium salt form of cefoperazone. It is a semi-synthetic, broad-spectrum, and third-generation cephalosporin compound with antibiotic and bactericidal activities. | CAS: 62893-20-3 | C25H29N9NaO8S2 | 670.672 g/mol

Supplier:  Thermo Scientific Chemicals J6518506

 View more versions of this product

Catalog No. AAJ6518506


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Cefoperazone sodium salt is a semi-synthetic, broad-spectrum, third-generation cephalosporin compound with antibiotic and bactericidal activities
• This compound can inhibit bacterial cell wall synthesis by inactivating penicillin-binding proteins and consequently interfering with the final transpeptidation step required for cross-linking peptidoglycan units, an important component of bacterial cell walls. This leads to a reduction in cell wall stability, causing cell lysis

Application

• Cefoperazone sodium salt exhibits activity against β-lactamase resistant microorganisms
• This compound is also used for studying the expression, binding, and inhibition of penicillin-binding proteins
• Cefoperazone is active in vitro against most gram-positive and gram-negative bacteria, including β-lactamase strains
• This compound has the ability to inhibit ∝-antitrypsin

Specifications

Specifications

Cefoperazone Sodium Salt
Powder
White
C25H29N9NaO8S2
4902135
Cefobid sodium salt; Cefazone sodium salt
IHWPDDMUNKOOFD-BYLXUVCXSA-N
(6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
131673981
Cefoperazone sodium salt
62893-20-3
5 g
MFCD07793331
14,1930
Soluble in water; Slightly soluble in alcohol.
[HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na]
670.672
667.65

Chemical Identifiers

62893-20-3
670.672
IHWPDDMUNKOOFD-BYLXUVCXSA-N
131673981
[HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na]
C25H29N9NaO8S2
MFCD07793331
Cefobid sodium salt; Cefazone sodium salt
(6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

H317-H334

EINECSNumber : 263-751-5

RTECSNumber : XI0374000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only