missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefoperazone sodium salt, MP Biomedicals™
Cefoperazone sodium salt USP, White powder
Supplier: MP Biomedicals Inc 0219969505
Description
- It is used to study drug-protein binding, expression and inhibition of penicillin-binding proteins (PBPs) during cell wall synthesis.
- Cefoperazone sodium salt is a broad spectrum third generation cephalosporin antibiotic that has similar antimicrobial activity to ceftazidime.
- It is active against Pseudomonoas aeruginosa and it has enhanced activity against Enterobacteriaceae and Bacteroides sp. in the presence of the beta-lactamase inhibitor sulbactam. Compared to cefotaxime, cefoperazone has greater susceptibility to hydrolysis by some beta-lactamases.
Specifications
| 62893-20-3 | |
| 4.5 to 6.5 (1 in 4) | |
| MFCD07793331 | |
| 4902135 | |
| Soluble in water (50mg/mL - clear, faint yellow solution). | |
| 667.65 |
| White | |
| C25H26N9NaO8S2 | |
| 5 g | |
| Cefobid, Cefoperazone sodium, CPZ, T-1551, [6R-[6.alpha.,7.beta.(R*)]]-7- [[[[(4-Ethyl-2,3- dioxo-1-piperazinyl) carbonyl]amino] (4-hydroxyphenyl) acetyl] amino]-3- [[(1-methyl-1H-tetrazol-5-yl) thio] methyl]- 8-oxo-5- thia-1-azabicyclo[4.2.0], oct-2-ene-2- carboxylic acid, 7-[D-(-)-alpha- (4-ethyl-2,3-dioxo- 1-piperazinecarboxamido)- alpha-(4-hydroxyphenyl) acetamido]- 3-[[(1-methyl-1H- tetrazol-5-yl) thio] methyl]-3-cephem-4- carboxylic acid | |
| NCFTXMQPRQZFMZ-WERGMSTESA-M | |
| Powder |
Chemical Identifiers
| 62893-20-3 | |
| 667.65 | |
| NCFTXMQPRQZFMZ-WERGMSTESA-M |
| C25H26N9NaO8S2 | |
| MFCD07793331 |
Safety and Handling
storageNote1 : +4°C