missing translation for 'onlineSavingsMsg'
Learn More

Cefmetazole Sodium Salt 92.0+%, TCI America™

Catalog No. c3029250mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
C3029250MG 250 mg
C30291G 1 g
2 options

Catalog No. C3029250MG

Supplier: TCI America C3029250MG

Only null left

Chemical Identifiers

CAS 56796-39-5
Molecular Formula C15H16N7NaO5S3
Molecular Weight (g/mol) 493.507
MDL Number MFCD00214260
InChI Key BITQGIOJQWZUPL-PBCQUBLHSA-M
Synonym cefmetazole sodium, zefazone, cefmetazon, cmz sodium, cefmetazole sodium salt, cs 1170 sodium, skf 83088 sodium, cefmetazone, unii-37y9vr4w7a
PubChem CID 23666711
ChEBI CHEBI:3490
IUPAC Name sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 250 mg
Formula Weight 493.51
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cefmetazole Sodium Salt
RTECSNumber XI0372200
TSCA No