missing translation for 'onlineSavingsMsg'
Learn More

Cefmetazole Sodium Salt 92.0+%, TCI America™

Supplier:  TCI America C30291G

 View more versions of this product

Catalog No. C30291G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cefmetazole Sodium Salt
White-Yellow
MFCD00214260
cefmetazole sodium, zefazone, cefmetazon, cmz sodium, cefmetazole sodium salt, cs 1170 sodium, skf 83088 sodium, cefmetazone, unii-37y9vr4w7a
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+]
493.507
CHEBI:3490
≥97.0% (T)
56796-39-5
C15H16N7NaO5S3
1 g
BITQGIOJQWZUPL-PBCQUBLHSA-M
sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
23666711
493.51
Crystalline Powder

Chemical Identifiers

56796-39-5
493.507
BITQGIOJQWZUPL-PBCQUBLHSA-M
23666711
sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
C15H16N7NaO5S3
MFCD00214260
cefmetazole sodium, zefazone, cefmetazon, cmz sodium, cefmetazole sodium salt, cs 1170 sodium, skf 83088 sodium, cefmetazone, unii-37y9vr4w7a
CHEBI:3490
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XI0372200

TSCA : No

SDS
Documents

Documents

Promotions

Promotions