missing translation for 'onlineSavingsMsg'
Learn More

Cefixime, 98%, Thermo Scientific Chemicals

Catalog No. AC459480010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459480010 1 g
AC459480050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459480010 Supplier Thermo Scientific Chemicals Supplier No. 459480010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79350-37-1
Molecular Formula C16H15N5O7S2
Molecular Weight (g/mol) 453.44
MDL Number MFCD00865020
InChI Key OKBVVJOGVLARMR-QSWIMTSFSA-N
Synonym cefixime, cefixima, cefiximum, cefixim, cefspan, cephoral, cefixoral, cefiximum latin, suprax, necopen
PubChem CID 5362065
ChEBI CHEBI:472657
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O

Specifications

pH 2.6 to 4.1
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 453.45
Percent Purity 98%
Chemical Name or Material Cefixime

RUO – Research Use Only