missing translation for 'onlineSavingsMsg'
Learn More

Cefepime hydrochloride monohydrate

Catalog No. p-7057469
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6623703 1 g
AAJ6623706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6623703 Supplier Thermo Scientific Chemicals Supplier No. J6623703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cefepime hydrochloride monohydrate, CAS # 123171-59-5, is a hydrochloride salt of the antibiotic derived from Acremonium fungus. | CAS: 123171-59-5 | C19H29Cl2N6O6S2+ | 572.497 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Cefepime hydrochloride is a fourth-generation cephalosporin antibiotic that works against most antibiotic resistant strains of bacteria

• Cefepime hydrochloride is a penicillin binding protein inhibitor that prevents peptidoglycan cross-linkage and leads to cell wall weakening and lysis

Application

• Cefepime hydrochloride monohydrate can be used for the laboratory study of microbial physiology and antibiotic resistance

• Cefepime has broad-spectrum activity; it is appropriate for research on bacteria responsible for resistant infections and physiological studies

Chemical Identifiers

CAS 123171-59-5
Molecular Formula C19H29Cl2N6O6S2+
Molecular Weight (g/mol) 572.497
MDL Number MFCD01687679
InChI Key LRAJHPGSGBRUJN-OMIVUECESA-O
PubChem CID 129893984
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
SMILES C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl

Specifications

Type Cefepime Hydrochloride Monohydrate
Physical Form Powder
Melting Point 150°C
Color White to Yellow
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in DMSO
Formula Weight 571.5
Chemical Name or Material Cefepime hydrochloride monohydrate
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only