missing translation for 'onlineSavingsMsg'
Learn More

Cefcapene Pivoxil Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. C2623200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2623200MG 200 mg
C26231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2623200MG Supplier TCI America Supplier No. C2623200MG
Only null left

Chemical Identifiers

CAS 147816-24-8
Molecular Formula C23H30ClN5O8S2
Molecular Weight (g/mol) 604.09
MDL Number MFCD01682041
InChI Key ALPFRUJYOAKQQR-CQZSJNSUSA-N
PubChem CID 6918126
IUPAC Name 2,2-dimethylpropanoyloxymethyl (6R,7R)-7-[[(Z)-2-(2-amino-1,3-thiazol-4-yl)pent-2-enoyl]amino]-3-(carbamoyloxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;hydrochloride
SMILES CCC=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)OCOC(=O)C(C)(C)C.Cl

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 604.09
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Cefcapene Pivoxil Hydrochloride Monohydrate
RTECSNumber XI0325000
TSCA No
Recommended Storage Refrigerator