accessibility menu, dialog, popup

UserName

H2OClH3COHN(R)(R)SNOOOOOCH3CH3H3CONH2OSNH2NH(Z)

CAS RN 147816-24-8

IUPAC Name: hydrogen [(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)pent-2-enamido]-3-[(carbamoyloxy)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy]methyl 2,2-dimethylpropanoate hydrate chloride
Synonyms: hydrogen [(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)pent-2-enamido]-3-[(carbamoyloxy)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy]methyl 2,2-dimethylpropanoate hydrate chloride
Molecular Weight (g/mol): 622.11
Molecular Formula: C23H32ClN5O9S2
InChi Key: LUXIJPQYUCFVAL-XRLCNELCSA-N
SMILES: [H+].O.[Cl-].CC\C=C(/C(=O)N[C@H]1[C@H]2SCC(COC(N)=O)=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)C1=CSC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results