missing translation for 'onlineSavingsMsg'
Learn More

Cefazolin sodium salt

Catalog No. AAJ6527403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6527403 1 g
AAJ6527406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6527403 Supplier Thermo Scientific Chemicals Supplier No. J6527403
Only null left

Inhibits bacterial cell wall synthesis | CAS: 27164-46-1 | C14H16N8NaO4S3 | 479.504 g/mol

Inhibits bacterial cell wall synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27164-46-1
Molecular Formula C14H16N8NaO4S3
Molecular Weight (g/mol) 479.504
MDL Number MFCD00056883
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
Synonym Cefamedin
PubChem CID 131673922
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]

Specifications

Type Cefazolin Sodium Salt
Physical Form Powder
Melting Point 170°C to 172°C
Color White
Quantity 1 g
Sensitivity Light sensitive
Beilstein 3585038
Merck Index 14,1917
Formula Weight 476.48
Chemical Name or Material Cefazolin sodium salt
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 248-278-4
RTECSNumber XI0390000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only