missing translation for 'onlineSavingsMsg'
Learn More

Cefazolin Sodium Salt 98.0+%, TCI America™

Catalog No. p-7125353
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22425G 5 g
C224225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22425G Supplier TCI America Supplier No. C22425G
Only null left

Chemical Identifiers

CAS 27164-46-1
Molecular Formula C14H16N8NaO4S3
Molecular Weight (g/mol) 479.504
MDL Number MFCD00056883
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
Synonym cefazolin sodium, cefazoline sodium, cefazolin sodium salt, sodium cefazolin, sodium cephazolin, kefzol, ancef, sodium cez, monosodium cefazolin
PubChem CID 131673922
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 5 g
Formula Weight 476.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cefazolin Sodium Salt
EINECSNumber (9)-2271
RTECSNumber XI0390000
TSCA No
Recommended Storage Refrigerator