missing translation for 'onlineSavingsMsg'
Learn More

Cefazolin Sodium Salt, MP Biomedicals™

Catalog No. p-4405067
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15494690 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15494690 Supplier MP Biomedicals Inc Supplier No. 0215494690
Only null left

Chemical Identifiers

CAS 27164-46-1
Molecular Formula C14H16N8NaO4S3
Molecular Weight (g/mol) 479.504
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
Synonym Acef, Cefazil
PubChem CID 131673922
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]

Specifications

Type Cefazolin Sodium Salt
Physical Form Powder
Color White
Quantity 500 mg
Formula Weight 476.48
Chemical Name or Material Cefazolin Sodium Salt