missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefadroxil, 95-105%

Catalog No. AC459470010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459470010 1 g
AC459470050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459470010 Supplier Thermo Scientific Chemicals Supplier No. 459470010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66592-87-8
Molecular Formula C16H17N3O5S
Molecular Weight (g/mol) 363.39
InChI Key BOEGTKLJZSQCCD-UEKVPHQBSA-N
Synonym cefadroxil, cephadroxil, cefadroxil anhydrous, cefadroxilo, cefadroxilum, d-cefadroxil, duricef, ultracef, cefadroxilum inn-latin, unii-q525pa8jjb
PubChem CID 47965
ChEBI CHEBI:3479
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O

Specifications

pH 4.0 to 6.0
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 363.39
Percent Purity 95 to 105%
Chemical Name or Material Cefadroxil
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only