accessibility menu, dialog, popup

UserName

CAS RN 66592-87-8

IUPAC Name: (6R,7R)-7-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
Synonyms: cefadroxil cefadroxil 1-wasser cefadroxil monohydrate
Molecular Weight (g/mol): 381.4
Molecular Formula: C16H19N3O6S
InChi Key: NBFNMSULHIODTC-CYJZLJNKSA-N
SMILES: O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=C(O)C=C1)C2=O)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results