missing translation for 'onlineSavingsMsg'
Learn More

Cefaclor Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015290
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015290 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015290 Supplier Merck Emd Millipore Supplier No. PHR12831G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Cefaclor belongs to the class of second generation semi-synthetic cephalosporin antibiotic drugs that is normally administered orally. With its broad-spectrum antibacterial action, cefaclor is effective against several gram-positive as well as gram-negative bacteria. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 70356-03-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C15H14 ClN3O4 S · H2O
Synonym Cefaclor monohydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 385.82
Grade Certified Reference Material