missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CE Phosphoramidite, dC(Bz)

The dC(Bz) CE Phosphoramidite is used for nucleic acid synthesis on ABI DNA Synthesizers. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.β-cyanoethyl phosphoramidites are for use on ABI synthesizers to guarantee you high quality nucleic acid synthesis.

Supplier:  Applied Biosystems™ 400328

Encompass

 View more versions of this product

Catalog No. 400328


Only null left

Description

Description

High QualityOur phosphoramidites benefit from proprietary manufacturing processes that yield high-purity products. In synthesis they perform at a correspondingly high level, producing pure, High Quality DNA at all scales. We guarantee that 5'-dimethoxytrityl nucleoside β-cyanoethyl phosphoramidites, monomer intermediates for licensed β-cyanoethyl phosphoramidite chemistry, will produce DNA that is at least 99.8% chemically authentic when used with recommendedsynthesis cycles.Dry phosphoroamidites (powder form) are guaranteed for one year from the date of shipment. Lifetime of diluted phosphoramidites in residence on the instrument is two weeks when used with CPG synthesis supports, and three weeks when used with polystyrene supports.
  • Proprietary manufacturing processes yield high-purity β-cyanoethyl phosphoramidites, which in turn guarantee production of high-quality DNA at all scales.
  • β-cyanoethyl phosphoramidites are use-tested and optimized for use on the ABI synthesizers to guarantee you high-quality nucleic acid synthesis.
Specifications

Chemical Identifiers

Store in freezer (-5 to -30°C).
3900 DNA Synthesizer
Deoxycytidine (dC)
Oligonucleotide Synthesis Reagent
Room Temperature
dC(Bz) CEP
DNA
Oligo Synthesis
DNA
Phosphoramidite
Powder

Specifications

dC(Bz) CEP
DNA
Oligo Synthesis
DNA
1 g
Room Temperature
Store in freezer (-5 to -30°C).
3900 DNA Synthesizer
Deoxycytidine (dC)
Oligonucleotide Synthesis Reagent
Phosphoramidite
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.