missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa559393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559393 Supplier Invitrogen™ Supplier No. PA559393
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: GKWEYLIRWK GYGSTEDTWE PEHHLLHCEE FIDEFNGLHM SKDKRIKSGK QSSTSKLLRD SRGPSVEKLS HRPSDPGKSK GTSHKRKRIN PPL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

CDYL2 (Chromodomain Y Like 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includemethylated histone binding. An important paralog of this gene isCDYL.

Specifications

Antigen CDYL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDYL2
Gene Accession No. Q8N8U2
Gene Alias 1700029M19Rik; 4930453I21Rik; Cdyl2; CDY-like 2; chromodomain protein, Y chromosome-like 2; chromodomain protein, Y-like 2; chromodomain Y-like protein 2; Gm20194; PCCP1; prostate cancer candidate protein 1, PCCP1
Gene Symbols CDYL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDYL2. Recombinant protein control fragment (Product #RP-98695).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less