missing translation for 'onlineSavingsMsg'
Learn More

CDTA (Free Acid), ACS Reagent (1,2-Cyclohexylenedinitrilo)tetraacetic Acid Monohydrate), Reagents

Catalog No. p-200021102
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338025 100 g
01338024 25 g
01338023 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338025 Supplier Labchem Inc Supplier No. C1194010100C2
Only null left

CDTA (Free Acid), ACS Reagent (1,2-Cyclohexylenedinitrilo)tetraacetic Acid Monohydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
Synonym trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid monohydrate
IUPAC Name 2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

Color Colorless/White
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.5
Grade ACS
Physical Form Solid
Chemical Name or Material CDTA (Free Acid), ACS Reagent (1,2-Cyclohexylenedinitrilo)tetraacetic Acid Monohydrate)