missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5 Recombinant Rabbit Monoclonal Antibody (23GB1425)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553157
Description
CDK5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain. Active form of CDK5, which has also been called neuronal cdc2-like kinase, is a heterodimer of CDK5 and a 25 kDa protein which is derived proteolytically from a 35 kDa brain and neuron-specific protein and is essential for the kinase activity of CDK5. CDK5 has emerged as a crucial regulator of neuronal migration in the developing central nervous system. CDK5 phosphorylates a diverse list of substrates, implicating it in the regulation of a range of cellular processes - from adhesion and motility, to synaptic plasticity and drug addiction.Chemical Identifiers
| CDK5 | |
| 1.6 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49615, Q00535, Q03114 | |
| 1020, 12568, 140908 | |
| A synthesized peptide derived from human Cdk5 (250-292AA). | |
| Antibody |
| 23GB1425 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CDK5 | |
| AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit | |
| CDK5 | |
| Primary | |
| Affinity chromatography |
Specifications
| CDK5 | |
| Recombinant Monoclonal | |
| 1.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49615, Q00535, Q03114 | |
| CDK5 | |
| A synthesized peptide derived from human Cdk5 (250-292AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1425 | |
| Unconjugated | |
| CDK5 | |
| AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1020, 12568, 140908 | |
| -20°C | |
| Liquid |