Learn More
Invitrogen™ Human CDK4/Cyclin D1, GST Tag Recombinant Protein
Recombinant Protein
$908.00 - $22370.00
Specifications
| Accession Number | P11802, P24385 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris with 0.02% Triton X-100, 150mM NaCl, 2mM DTT, 0.5mM EDTA, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 1019, 595 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR8064A
|
Invitrogen™
PR8064A |
100 μg |
Each for $4,740.00
|
|
|||||
PV4400
|
Invitrogen™
PV4400 |
10 μg |
Each for $908.00
|
|
|||||
PV4436
|
Invitrogen™
PV4436 |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.
Cyclin D1 and CDK4 (Cyclin-Dependent Kinase 4) are integral components of the cell cycle regulatory machinery, specifically involved in the transition from the G1 phase to the S phase. Cyclin D1 forms a complex with CDK4, enabling the phosphorylation of the retinoblastoma protein (pRB), which in turn releases E2F transcription factors, promoting the transcription of genes necessary for DNA synthesis and cell cycle progression. This Cyclin D1-CDK4 complex is localized both in the nucleus and cytoplasm, influenced by its binding partners, and Cyclin D1 can also be associated with the outer mitochondrial membrane and plasma membrane, indicating its role in extranuclear signaling and function. This complex is not only imperative for regulating the cell cycle but also plays a crucial role in glucose metabolism, as observed in diabetic models where Cyclin D1-CDK4 activity normalizes glycemia independently of insulin-mediated cell division. Dysregulation of the Cyclin D1-CDK4 pathway is often associated with cancer, as overexpression or amplification of Cyclin D1 can lead to uncontrolled cell proliferation observed in various malignancies such as breast, esophageal, and head and neck cancers.Specifications
| P11802, P24385 | |
| Kinase Assay | |
| 1019, 595 | |
| Human CDK4/Cyclin D1, GST Tag | |
| Purified | |
| RUO | |
| CDK4/Cyclin D1 | |
| CDK and CHK Kinase Family | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| SerâThr Kinases: CMCG Group | |
| Baculovirus | |
| Recombinant, Full Length |
| See Label | |
| 50mM tris with 0.02% Triton X-100, 150mM NaCl, 2mM DTT, 0.5mM EDTA, 50% glycerol and no preservative; pH 7.5 | |
| 61.8-61.2 kDa | |
| 7.5 | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903) | |
| CCND1, CDK4 | |
| Protein | |
| Human | |
| GST-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Dry Ice | |
| Kinases & Inhibitors | |
| Liquid |