missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK4/Cyclin D1, GST Tag Recombinant Protein

Catalog No. PR8064A
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR8064A 100 μg
PV4400 10 μg
PV4436 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR8064A Supplier Invitrogen™ Supplier No. PR8064A

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

Cyclin D1 and CDK4 (Cyclin-Dependent Kinase 4) are integral components of the cell cycle regulatory machinery, specifically involved in the transition from the G1 phase to the S phase. Cyclin D1 forms a complex with CDK4, enabling the phosphorylation of the retinoblastoma protein (pRB), which in turn releases E2F transcription factors, promoting the transcription of genes necessary for DNA synthesis and cell cycle progression. This Cyclin D1-CDK4 complex is localized both in the nucleus and cytoplasm, influenced by its binding partners, and Cyclin D1 can also be associated with the outer mitochondrial membrane and plasma membrane, indicating its role in extranuclear signaling and function. This complex is not only imperative for regulating the cell cycle but also plays a crucial role in glucose metabolism, as observed in diabetic models where Cyclin D1-CDK4 activity normalizes glycemia independently of insulin-mediated cell division. Dysregulation of the Cyclin D1-CDK4 pathway is often associated with cancer, as overexpression or amplification of Cyclin D1 can lead to uncontrolled cell proliferation observed in various malignancies such as breast, esophageal, and head and neck cancers.

Specifications

Accession Number P11802, P24385
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 150mM NaCl, 2mM DTT, 0.5mM EDTA, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1019, 595
Molecular Weight (g/mol) 61.8-61.2 kDa
Name Human CDK4/Cyclin D1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903)
Common Name CDK4/Cyclin D1
Gene Symbol CCND1, CDK4
Kinase Family CDK and CHK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
Target Entry CDK4/Cyclin D1
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Cell Cycle & Proliferation, Signal Transduction
Show More Show Less