Learn More
Invitrogen™ Human CDK11 (Inactive), GST Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PV6284
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human CDK11 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for CDK11 (Inactive) is NP_055891.1.
CDC2L6 is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.Specifications
| Q9BWU1 | |
| Kinase Assay | |
| 23097 | |
| Human CDK11 (Inactive), GST Tag | |
| 2700084L06Rik; AI845279; AW228747; bA346C16.3; Cdc2l6; CDC2-related protein kinase 6; Cdk11; Cdk19; CDK8-like cyclin-dependent kinase; cell division cycle 2-like 6 (CDK8-like); cell division cycle 2-like protein kinase 6; Cell division protein kinase 19; cyclin dependent kinase 19; cyclin dependent kinase 19 variant 2; cyclin-dependent kinase (CDC2-like) 11; cyclin-dependent kinase 11; Cyclin-dependent kinase 19; death-preventing kinase; KIAA1028; mKIAA1028 | |
| 1-795 | |
| Human | |
| GST-tag | |
| Baculovirus |
| See Label | |
| 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
| 84.2 kDa | |
| 100 μg | |
| CDC2L6 | |
| Unconjugated | |
| Recombinant | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Liquid |