missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDH4 Recombinant Rabbit Monoclonal Antibody (23GB5055)
GREENER_CHOICE

Catalog No. MA552776
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552776 100 μL
MA552777 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552776 Supplier Invitrogen™ Supplier No. MA552776
Only null left

Rabbit Recombinant Monoclonal Antibody

CDH4 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CDH4 is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Based on studies in chicken and mouse, this cadherin is thought to play an important role during brain segmentation and neuronal outgrowth. In addition, a role in kidney and muscle development is indicated. Of particular interest are studies showing stable cis-heterodimers of cadherins 2 and 4 in cotransfected cell lines. Previously thought to interact in an exclusively homophilic manner, this is the first evidence of cadherin heterodimerization. Three transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen CDH4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5055
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cdh4
Gene Accession No. P39038, P55283
Gene Alias AW120700; CAD4; Cadherin; cadherin 4; cadherin 4 type 1 R-cadherin (retinal); cadherin 4, type 1, preproprotein; cadherin 4, type 1, R-cadherin (retinal); cadherin-4; Cdh4; Cdh4l; Rcad; R-CAD; R-Cadh; R-cadherin; retina cadherin; retinal cadherin
Gene Symbols Cdh4
Host Species Rabbit
Immunogen A synthesized peptide derived from human R Cadherin (800-900AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1002, 12561
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less