missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDH10 Recombinant Rabbit Monoclonal Antibody (23GB1600)

Catalog No. MA552497
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552497 20 μL
MA552496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552497 Supplier Invitrogen™ Supplier No. MA552497
Only null left

Rabbit Recombinant Monoclonal Antibody

CDH10 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CDH10 is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is predominantly expressed in brain and is putatively involved in synaptic adhesions, axon outgrowth and guidance.

Specifications

Antigen CDH10
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1600
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cdh10
Gene Accession No. P70408, Q9Y6N8
Gene Alias A830016G23Rik; C030003B10Rik; C030011H18Rik; Cadherin; cadherin 10; cadherin 10 type 2; cadherin 10, type 2 (T2-cadherin); cadherin-10; Cdh10; T2-cadherin
Gene Symbols Cdh10
Host Species Rabbit
Immunogen A synthesized peptide derived from human Cadherin 10 (750-788AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1008, 320873
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less